Thiophene-2-carbaldehyde ≥98.0% stabilized

Supplier: TCI America
Warning

T0725-25ML T0725-500ML T0725-100ML
TCT0725-025MLEA 102.7 CAD
TCT0725-025ML TCT0725-500ML TCT0725-100ML
Thiophene-2-carbaldehyde ≥98.0% stabilized
Thiophene-2-carbaldehyde

(stabilized with HQ)
CAS Number: 98-03-3
MDL Number: MFCD00005429
Molecular Formula: C5H4OS
Molecular Weight: 112.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 198
Flash Point (°C): 78
Specific Gravity (20/20): 1.23
Storage Temperature: 0-10°C

Formula: C₅H₄OS
Boiling Pt: 198 °C
Melting Pt: ? 10 °C
Density: 1.23 (20 °C)
Flash Pt: 78 °C
MDL Number: MFCD00005429
CAS Number: 98-03-3
EINECS: 202-629-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=330bc0fa1bf25c05ef226d85dbc43e7f0afd9862&vid=330bc0fa1bf25c05ef226d85dbc43e7f0afd9862&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR