2-Methylhippuric acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
T0719-5G
T0719-25G
TCT0719-005GEA
148.84
CAD
TCT0719-005G
TCT0719-025G
2-Methylhippuric acid ≥98.0% (by HPLC, titration analysis)
2-Methylhippuric acid
CAS Number: 42013-20-7
MDL Number: MFCD00050991
Molecular Formula: C10H11NO3
Molecular Weight: 193.20
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 163
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 2-Methylhippuric Acid, N-(2-Methylbenzoyl)glycine
Formula:
C₁₀H₁₁NO₃ Melting Pt: 163 °C |
MDL Number:
MFCD00050991 CAS Number: 42013-20-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...