1,14-Tetradecanedioic acid ≥98.0% for synthesis

Supplier: TCI America
Warning

Synonyms: Tetradecanedioic acid, 1,12-Dodecanedicarboxylic acid

T0704-25G T0704-5G
TCT0704-025GEA 272.39 CAD
TCT0704-025G TCT0704-005G
1,14-Tetradecanedioic acid ≥98.0% for synthesis
1,14-Tetradecanedioic acid

CAS Number: 821-38-5
MDL Number: MFCD00002743
Molecular Formula: C14H26O4
Molecular Weight: 258.36
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 127


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,12-Dodecanedicarboxylic Acid

Formula: C₁₄H₂₆O₄
MW: 258.36 g/mol
Boiling Pt: 407
Melting Pt: 126
Density: 1.108 g/cm³ (21 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002743
CAS Number: 821-38-5
EINECS: 212-476-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5746f68ff26208921f3b11fe5db0d3a429e3e97d&vid=5746f68ff26208921f3b11fe5db0d3a429e3e97d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR