n-Tetratriacontane ≥95.0% for synthesis

Supplier: TCI America

Synonyms: Tetratriacontane

T0687-25G T0687-1G T0687-5G
TCT0687-025GEA 3272.32 CAD
TCT0687-025G TCT0687-001G TCT0687-5G
n-Tetratriacontane ≥95.0% for synthesis
n-Tetratriacontane

CAS Number: 14167-59-0
MDL Number: MFCD00009412
Molecular Formula: C34H70
Molecular Weight: 478.93
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 285
Melting point (°C): 73

Formula: H₃C(CH₂)₃₂CH₃
MW: 478.93 g/mol
Boiling Pt: 285 °C
Melting Pt: 73 °C
Density: 0.7728 g/cm³ (90 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009412
CAS Number: 14167-59-0
EINECS: 238-013-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7a89fd75f3537757b3c4bc1892481864829e599&vid=7c0ea9a8d0cffe2149d49b2b9a42cd18e074daa9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR