Tricine ≥99.0% for synthesis

Supplier: TCI America

Synonyms: 2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine

T0682-25G T0682-250G
TCT0682-025GEA 56.28 CAD
TCT0682-025G TCT0682-250G
Tricine ≥99.0% for synthesis
Tricine

[Good's buffer component for biological research]
CAS Number: 5704-04-1
MDL Number: MFCD00004277
Molecular Formula: C6H13NO5
Molecular Weight: 179.17
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Color: White
Flash Point (°C): -21


Product Categories: Bioscience, Good's Buffering Agents
Other Product Names / Synonyms: N-[Tris(hydroxymethyl)methyl]glycine

Formula: (HOCH₂)₃CNHCH₂CO₂H
MW: 179.17 g/mol
Melting Pt: 187
Density: 1.523 g/cm³ (19.85 °C)
Flash Pt: –21 °C
Storage Temperature: Ambient
MDL Number: MFCD00004277
CAS Number: 5704-04-1
EINECS: 227-193-6
Merck Index: 13,09724
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66a1bbba6c838b918da6a8aca9f5c25fb4a73a66&vid=63c4c67704ef3a1a4ab1a87e299da20d212eaecb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR