Tris(2-carboxyethyl)isocyanurate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
3,3',3''-(2,4,6-Trioxo-1,3,5-triazinane-1,3,5-triyl)tripropionic acid, Isocyanuric Acid Tris(2-carboxyethyl) Ester
T0675-25G
T0675-500G
TCT0675-025GEA
56.88
CAD
TCT0675-025G
TCT0675-500G
Tris(2-carboxyethyl)isocyanurate ≥98.0% (by titrimetric analysis)
Tris(2-carboxyethyl)isocyanurate
CAS Number: 2904-41-8
MDL Number: MFCD00059790
Molecular Formula: C12H15N3O9
Molecular Weight: 345.26
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 228
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Isocyanuric Acid Tris(2-carboxyethyl) Ester
Formula:
C₁₂H₁₅N₃O₉ MW: 345.26 g/mol Melting Pt: 228 °C |
MDL Number:
MFCD00059790 CAS Number: 2904-41-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...