Tripropylene glycol monomethyl ether (mixture of isomer) ≥93.0%
Supplier: TCI America
T0651-25G
T0651-500G
TCT0651-025GEA
28.95
CAD
TCT0651-025G
TCT0651-500G
Tripropylene glycol monomethyl ether (mixture of isomer) ≥93.0%
Tripropylene glycol monomethyl ether (mixture of isomer)
CAS Number: 25498-49-1
MDL Number: MFCD00059142
Molecular Formula: C10H22O4
Molecular Weight: 206.28
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 243
Melting point (°C): -79
Flash Point (°C): 118
Specific Gravity (20/20): 0.97
Formula:
C₁₀H₂₂O₄ Boiling Pt: 243 °C Melting Pt: –79 °C Density: 0.97 (20 °C) Flash Pt: 118 °C |
MDL Number:
MFCD00059142 CAS Number: 25498-49-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...