Tetrabutylthiuram disulfide ≥98.0%

Supplier: TCI America

Synonyms: Bis(dibutylthiocarbamoyl) Disulfide, Bis(dibutylthiocarbamoyl) Disulphide

T0635-25G T0635-500G
TCT0635-025GEA 71.1 CAD
TCT0635-025G TCT0635-500G
Tetrabutylthiuram disulfide ≥98.0%
Tetrabutylthiuram disulfide

CAS Number: 1634-02-2
MDL Number: MFCD00027191
Molecular Formula: C18H36N2S4
Molecular Weight: 408.74
Purity/Analysis Method: >98.0% (N)
Form: Clear Liquid
Color: Reddish Yellow
Flash Point (°C): 138
Specific Gravity (20/20): 1.06


Product Categories: Synthetic Organic Chemistry, Thiocarbonyl Compounds
Other Product Names / Synonyms: Bis(dibutylthiocarbamoyl) Disulfide

Formula: C₁₈H₃₆N₂S₄
Density: 1.06 (20 °C)
Flash Pt: 138 °C
MDL Number: MFCD00027191
CAS Number: 1634-02-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ccf1384e381fa1cb0a1bde8603129916a37bcd6&vid=0ccf1384e381fa1cb0a1bde8603129916a37bcd6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR