3,3,5-Trimethylcaproic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T0630-25ML T0630-500ML
TCT0630-025MLEA 44.24 CAD
TCT0630-025ML TCT0630-500ML
3,3,5-Trimethylcaproic acid ≥98.0% (by titrimetric analysis)
3,3,5-Trimethylcaproic acid

CAS Number: 3302-10-1
MDL Number: MFCD00020507
Molecular Formula: C9H18O2
Molecular Weight: 158.24
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 120
Melting point (°C): -70
Flash Point (°C): 125
Specific Gravity (20/20): 0.90


Other Product Names / Synonyms: Isononanoic Acid

Formula: C₉H₁₈O₂
Boiling Pt: 120 °C
Melting Pt: –70 °C
Density: 0.90 (20 °C)
Flash Pt: 125 °C
MDL Number: MFCD00020507
CAS Number: 3302-10-1
EINECS: 221-975-0
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d8d6d07e4af72d1ed4975644f9da3552995fc82&vid=3d8d6d07e4af72d1ed4975644f9da3552995fc82&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR