2-Amino-6-methoxypyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2228-1G A2228-5G
TCA2228-1GEA 37.92 CAD
TCA2228-1G TCA2228-5G
2-Amino-6-methoxypyridine ≥98.0% (by GC, titration analysis)
2-Amino-6-methoxypyridine

CAS Number: 17920-35-3
MDL Number: MFCD04972542
Molecular Formula: C6H8N2O
Molecular Weight: 124.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 115
Specific Gravity (20/20): 1.15
Storage Temperature: 0-10°C

Formula: C₆H₈N₂O
MW: 124.14 g/mol
Boiling Pt: 115 °C
Density: 1.15 (20 °C)
MDL Number: MFCD04972542
CAS Number: 17920-35-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81c029429caf5ef717ddcf641f97143111a74ec1&vid=81c029429caf5ef717ddcf641f97143111a74ec1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR