2,4,6-Tribromobenzoic acid ≥98.0%
Supplier: TCI America
T0619-1G
T0619-5G
TCT0619-001GEA
201.77
CAD
TCT0619-001G
TCT0619-5G
2,4,6-Tribromobenzoic acid ≥98.0%
2,4,6-Tribromobenzoic acid
CAS Number: 633-12-5
MDL Number: MFCD00059882
Molecular Formula: C7H3Br3O2
Molecular Weight: 358.81
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 192
Formula:
C₇H₃Br₃O₂ Melting Pt: 192 °C |
MDL Number:
MFCD00059882 CAS Number: 633-12-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...