2-Ethylhexyl mercaptoacetate ≥98.0%

Supplier: TCI America

T0612-500ML T0612-25ML
TCT0612-500MLEA 137.46 CAD
TCT0612-500ML TCT0612-025ML
2-Ethylhexyl mercaptoacetate ≥98.0%
2-Ethylhexyl mercaptoacetate

CAS Number: 7659-86-1
MDL Number: MFCD00022081
Molecular Formula: C10H20O2S
Molecular Weight: 204.33
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 136
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: Thioglycolic Acid 2-Ethylhexyl Ester, Mercaptoacetic Acid 2-Ethylhexyl Ester, Octyl Thioglycolate, Thioglycolic Acid Octyl Ester, 2-Ethylhexyl Mercaptoacetate

Formula: C₁₀H₂₀O₂S
Density: 0.97 (20 °C)
Flash Pt: 136 °C
MDL Number: MFCD00022081
CAS Number: 7659-86-1
EINECS: 231-626-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2953ffd3c98240810665a7e4dac1a87005b14b39&vid=2953ffd3c98240810665a7e4dac1a87005b14b39&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR