Bis(2-ethylhexyl)-4-cyclohexene-1,2-dicarboxylate ≥97.0%
Supplier: TCI America
Synonyms:
4-Cyclohexene-1,2-dicarboxylic Acid Di(2-ethylhexyl) Ester, Dioctyl 4-Cyclohexene-1,2-dicarboxylate, 4-Cyclohexene-1,2-dicarboxylic Acid Bis(2-ethylhexyl) Ester
CAS Number: 2915-49-3
MDL Number: MFCD00059565
Molecular Formula: C24H42O4
Molecular Weight: 394.60
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 202
Specific Gravity (20/20): 0.97
Other Product Names / Synonyms: 4-Cyclohexene-1,2-dicarboxylic Acid Di(2-ethylhexyl) Ester, Dioctyl 4-Cyclohexene-1,2-dicarboxylate, 4-Cyclohexene-1,2-dicarboxylic Acid Bis(2-ethylhexyl) Ester
Formula:
C₂₄H₄₂O₄ Density: 0.97 (20 °C) Flash Pt: 202 °C |
MDL Number:
MFCD00059565 CAS Number: 2915-49-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...