2-Amino-5-chloro-4-methylbenzenesulfonic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
A0287-25G
A0287-500G
TCA0287-025GEA
44.24
CAD
TCA0287-025G
TCA0287-500G
2-Amino-5-chloro-4-methylbenzenesulfonic acid ≥98.0% (by HPLC, titration analysis)
2-Amino-5-chloro-4-methylbenzenesulphonic acid
CAS Number: 88-53-9
MDL Number: MFCD00043924
Molecular Formula: C7H8ClNO3S
Molecular Weight: 221.66
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Other Product Names / Synonyms: 4-Chloro-5-methylaniline-2-sulfonic Acid, 4-Chloro-m-toluidine-6-sulfonic Acid, 2-Amino-5-chloro-4-methylbenzenesulfonic Acid
Formula:
C₇H₈ClNO₃S |
MDL Number:
MFCD00043924 CAS Number: 88-53-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...