L-Tyrosine ethyl ester ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T0551-5G T0551-1G
TCT0551-005GEA 178.54 CAD
TCT0551-005G TCT0551-001G
L-Tyrosine ethyl ester ≥98.0% (by titrimetric analysis)
L-Tyrosine ethyl ester

CAS Number: 949-67-7
MDL Number: MFCD00063046
Molecular Formula: C11H15NO3
Molecular Weight: 209.25
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 103
Specific rotation [a]20/D: 17.5 deg (C=5, EtOH)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Tyr-OEt

Formula: C₁₁H₁₅NO₃
Melting Pt: 103 °C
MDL Number: MFCD00063046
CAS Number: 949-67-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=629a06429f927fffcf7d161a116b3dff274e567f&vid=629a06429f927fffcf7d161a116b3dff274e567f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR