Tween® 85 (Polysorbate)

Supplier: TCI America

Synonyms: Polyoxyethylenesorbitan Trioleate

T0547-500G T0547-25G
TCT0547-500GEA 67.21 CAD
TCT0547-500G TCT0547-025G
Tween® 85 (Polysorbate)
Tween® 85 (Polysorbate)

CAS Number: 9005-70-3
MDL Number: MFCD01779641
Form: Clear Liquid
Color: Yellow
Melting point (°C): -20
Flash Point (°C): 315
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: Polyoxyethylene Sorbitan Trioleate

Melting Pt: –20 °C
Density: 1.03 (20 °C)
Flash Pt: 315 °C
Storage Temperature: Ambient
MDL Number: MFCD01779641
CAS Number: 9005-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR