DL-Tropic acid ≥98.0%

Supplier: TCI America

Synonyms: 3-Hydroxy-2-phenylpropanoic acid, Tropic acid, 3-Hydroxy-2-phenylpropionic acid, (±)-Tropic acid, 2-Phenylhydracrylic acid

T0533-25G T0533-250G
TCT0533-025GEA 98.07 CAD
TCT0533-025G TCT0533-250G
DL-Tropic acid ≥98.0%
DL-Tropic acid

CAS Number: 552-63-6
MDL Number: MFCD00004255
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 118

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Melting Pt: 118 °C
MDL Number: MFCD00004255
CAS Number: 529-64-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e46135a66941952f1a5128f424ac54e4d0c80d97&vid=e46135a66941952f1a5128f424ac54e4d0c80d97&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR