Trilauryl trithiophosphite
Supplier: TCI America
Synonyms:
Tridodecyl Phosphonotrithioite, Phosphorotrithious Acid Tridodecyl Ester, Tridodecyl Trithiophosphite
T0460-25G
T0460-100G
TCT0460-025GEA
56.88
CAD
TCT0460-025G
TCT0460-100G
Trilauryl trithiophosphite
Trilauryl trithiophosphite
CAS Number: 1656-63-9
MDL Number: MFCD00015072
Molecular Formula: C36H75PS3
Molecular Weight: 635.15
Form: Crystal
Color: White
Flash Point (°C): 209
Other Product Names / Synonyms: Tridodecyl Phosphonotrithioite, Tridodecyl Trithiophosphite, Phosphorotrithious Acid Tridodecyl Ester
Formula:
C₃₆H₇₅PS₃ MW: 635.15 g/mol Flash Pt: 209 °C |
MDL Number:
MFCD00015072 CAS Number: 1656-63-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...