1,3,3-Trimethylindolino-β-naphthopyrylospiran ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1',3',3'-Trimethylspiro[benzo[f]chromene-3,2'-indoline], 1,3,3-Trimethylspiro[indoline-2,3'-[3H]naphth[2,1-b]pyran]

T0423-5G T0423-1G
TCT0423-005GEA 268.6 CAD
TCT0423-005G TCT0423-001G
1,3,3-Trimethylindolino-β-naphthopyrylospiran ≥98.0% (by titrimetric analysis)
1,3,3-Trimethylindolino-beta-naphthopyrylospiran

[Photochromic Compound]
CAS Number: 1592-43-4
MDL Number: MFCD00059909
Molecular Formula: C23H21NO
Molecular Weight: 327.43
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 174


Product Categories: Materials Chemistry, Photochromic Compounds
Other Product Names / Synonyms: 1,3,3-Trimethylspiro[indoline-2,3'-[3H]naphth[2,1-b]pyran]

Formula: C₂₃H₂₁NO
MW: 327.43 g/mol
Melting Pt: 174 °C
MDL Number: MFCD00059909
CAS Number: 1592-43-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ed05754ee7719b29950945cfc92d821f7b17eff&vid=2ed05754ee7719b29950945cfc92d821f7b17eff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR