Triisodecyl phosphite (mixture of isomers)

Supplier: TCI America

Synonyms: Phosphorous Acid Triisodecyl Ester

T0420-500ML T0420-25ML
TCT0420-500MLEA 110.6 CAD
TCT0420-500ML TCT0420-025ML
Triisodecyl phosphite (mixture of isomers)
Triisodecyl phosphite (mixture of isomers)

CAS Number: 25448-25-3
MDL Number: MFCD00015059
Molecular Formula: C30H63O3P
Molecular Weight: 502.80
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: Phosphorous Acid Triisodecyl Ester

Formula: C₃₀H₆₃O₃P
Density: 0.89 (20 °C)
MDL Number: MFCD00015059
CAS Number: 25448-25-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e299a8745a600e91e591344dfc0694875578abd1&vid=e299a8745a600e91e591344dfc0694875578abd1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR