Tris(decyl)amine ≥97.0% (by titrimetric analysis)
Supplier: TCI America
T0418-5G
T0418-25G
TCT0418-005GEA
93.22
CAD
TCT0418-005G
TCT0418-025G
Tris(decyl)amine ≥97.0% (by titrimetric analysis)
Tris(decyl)amine
CAS Number: 1070-01-5
MDL Number: MFCD00041914
Molecular Formula: C30H63N
Molecular Weight: 437.84
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 430
Flash Point (°C): 110
Specific Gravity (20/20): 0.82
Formula:
C₃₀H₆₃N Boiling Pt: 430 °C Melting Pt: –2 °C Density: 0.82 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00041914 CAS Number: 1070-01-5 EINECS: 213-966-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...