Tridecanol (mixture of isomers)

Supplier: TCI America

T0414-25ML T0414-500ML
TCT0414-025MLEA 57.84 CAD
TCT0414-025ML TCT0414-500ML
Tridecanol (mixture of isomers)
Tridecanol (mixture of isomers)

CAS Number: 26248-42-0
MDL Number: MFCD01778419
Molecular Formula: C13H28O
Molecular Weight: 200.37
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 130
Specific Gravity (20/20): 0.84


Other Product Names / Synonyms: Tridecyl Alcohol

Formula: C₁₃H₂₈O
Density: 0.84 (20 °C)
Flash Pt: 130 °C
MDL Number: MFCD01778419
CAS Number: 26248-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR