Tricaprin ≥98.0%

Supplier: TCI America

Synonyms: Tridecanoin, Trisdecanoin, 1,2,3-Tridecanoylglycerol, Glycerol Tridecanoate, Propane-1,2,3-triyl tris(decanoate)

T0413-25G T0413-10G
TCT0413-025GEA 327.99 CAD
TCT0413-025G TCT0413-010G
Tricaprin ≥98.0%
Tricaprin

CAS Number: 621-71-6
MDL Number: MFCD00036239
Molecular Formula: C33H62O6
Molecular Weight: 554.85
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 254
Melting point (°C): 32
Flash Point (°C): 76
Storage Temperature: 0-10°C


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Glycerol Tridecanoate, Tridecanoin, 1,2,3-Tridecanoylglycerol

Formula: C₃₃H₆₂O₆
Boiling Pt: 254 °C
Melting Pt: 32 °C
Flash Pt: 76 °C
MDL Number: MFCD00036239
CAS Number: 621-71-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8683bd58a00e113164301faaa32e0ac23fdd6fdf&vid=8683bd58a00e113164301faaa32e0ac23fdd6fdf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR