1,3,3-Trimethylindolino-6'-nitrobenzopyrylospiran ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
1',3',3'-Trimethyl-6-nitrospiro[chromene-2,2'-indoline], 1',3',3'-Trimethyl-6-nitrospiro[1(2H)-benzopyran-2,2'-indoline]
[Photochromic Compound]
CAS Number: 1498-88-0
MDL Number: MFCD00059175
Molecular Formula: C19H18N2O3
Molecular Weight: 322.36
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 178
Product Categories: Materials Chemistry, Photochromic Compounds
Other Product Names / Synonyms: 1',3',3'-Trimethyl-6-nitrospiro[1(2H)-benzopyran-2,2'-indoline]
Formula:
C₁₉H₁₈N₂O₃ MW: 322.36 g/mol Melting Pt: 178 °C |
MDL Number:
MFCD00059175 CAS Number: 1498-88-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...