1,3,3-Trimethylindolinobenzopyrylospiran ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 1',3',3'-Trimethylspiro[1(2H)-benzopyran-2,2'-indoline]

T0344-1G T0344-5G
TCT0344-001GEA 60.04 CAD
TCT0344-001G TCT0344-5G
1,3,3-Trimethylindolinobenzopyrylospiran ≥98.0% (by HPLC, titration analysis)
1,3,3-Trimethylindolinobenzopyrylospiran

[Photochromic Compound]
CAS Number: 1485-92-3
MDL Number: MFCD00059174
Molecular Formula: C19H19NO
Molecular Weight: 277.37
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 92


Product Categories: Materials Chemistry, Photochromic Compounds
Other Product Names / Synonyms: 1',3',3'-Trimethylspiro[1(2H)-benzopyran-2,2'-indoline]

Formula: C₁₉H₁₉NO
MW: 277.37 g/mol
Melting Pt: 92 °C
MDL Number: MFCD00059174
CAS Number: 1485-92-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fad71257ca1cf8640e3a533f029467d2ea097423&vid=fad71257ca1cf8640e3a533f029467d2ea097423&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR