Nɑ-Tosyl-L-arginine methyl ester hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: TAME, Nɑ-p-Toluenesulfonyl-L-arginine Methyl Ester Hydrochloride, Nα-Tosyl-L-arginine Methyl Ester HCl, Nɑ-p-Toluenesulphonyl-L-arginine Methyl Ester Hydrochloride, Tos-Arg-OMe.HCl, Nɑ-4-Tosyl-L-arginine methyl ester hydrochloride

T0330-25G T0330-1G
TCT0330-025GEA 328.64 CAD
TCT0330-025G TCT0330-001G
Nɑ-Tosyl-L-arginine methyl ester hydrochloride ≥98.0% (by titrimetric analysis)
Nalpha-Tosyl-L-arginine methyl ester hydrochloride

CAS Number: 1784-03-8
MDL Number: MFCD00012578
Molecular Formula: C14H22N4O4S
Molecular Weight: 378.87
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 147
Specific rotation [a]20/D: -14 deg (C=4, H2O)


Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: TAME, Nalpha-p-Toluenesulfonyl-L-arginine Methyl Ester Hydrochloride, Tos-Arg-OMe.HCl

Formula: C₁₄H₂₂N₄O₄S
MW: 378.87 g/mol
Melting Pt: 147 °C
MDL Number: MFCD00012578
CAS Number: 1784-03-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2016a79bb5d878777fe14a347ab5c5a786007634&vid=2016a79bb5d878777fe14a347ab5c5a786007634&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR