m-Tolyltetrazolium red ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T0324-100MG T0324-1G
TCT0324-100MGEA 51.19 CAD
TCT0324-100MG TCT0324-001G
m-Tolyltetrazolium red ≥98.0% (by titrimetric analysis)
m-Tolyltetrazolium red

CAS Number: 88159-25-5
MDL Number: MFCD00059997
Molecular Formula: C20H17ClN4
Molecular Weight: 348.83
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Very Pale Yellow


Other Product Names / Synonyms: 2,5-Diphenyl-3-(m-tolyl)tetrazolium Chloride

Formula: C₂₀H₁₇ClN₄
MDL Number: MFCD00059997
CAS Number: 88159-25-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3325be6031c5cf1687d161528df85950707cae83&vid=3325be6031c5cf1687d161528df85950707cae83&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR