m-Tolyldiethanolamine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2,2'-(3-Methylphenylimino)diethanol, 2,2'-(m-Tolylimino)diethanol, N,N-Bis(2-hydroxyethyl)-m-toluidine, 2,2'-(m-Tolylazanediyl)diethanol

T0315-25G T0315-500G
TCT0315-025GEA 53.72 CAD
TCT0315-025G TCT0315-500G
m-Tolyldiethanolamine ≥98.0% (by titrimetric analysis)
m-Tolyldiethanolamine

CAS Number: 91-99-6
MDL Number: MFCD00020578
Molecular Formula: C11H17NO2
Molecular Weight: 195.26
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 70


Other Product Names / Synonyms: N,N-Bis(2-hydroxyethyl)-m-toluidine, 2,2'-(3-Methylphenylimino)diethanol, 2,2'-(m-Tolylimino)diethanol

Formula: C₁₁H₁₇NO₂
MW: 195.26 g/mol
Melting Pt: 70 °C
MDL Number: MFCD00020578
CAS Number: 91-99-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65629cb0191b5499f7511e079dd95fbcb5a979e5&vid=65629cb0191b5499f7511e079dd95fbcb5a979e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR