1-(o-Tolyl)biguanide ≥97.0% (by titrimetric analysis)

Supplier: TCI America

T0314-25G T0314-500G
TCT0314-025GEA 90.06 CAD
TCT0314-025G TCT0314-500G
1-(o-Tolyl)biguanide ≥97.0% (by titrimetric analysis)
1-(o-Tolyl)biguanide

CAS Number: 93-69-6
MDL Number: MFCD00019731
Molecular Formula: C9H13N5
Molecular Weight: 191.24
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Melting point (°C): 147

Formula: C₉H₁₃N₅
Melting Pt: 147 °C
MDL Number: MFCD00019731
CAS Number: 93-69-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37de5a732cea475cc6b85cc74779aea134483ca2&vid=37de5a732cea475cc6b85cc74779aea134483ca2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR