3-Methylhippuric acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
T0312-25G
T0312-5G
TCT0312-025GEA
300.83
CAD
TCT0312-025G
TCT0312-5G
3-Methylhippuric acid ≥98.0% (by titrimetric analysis)
3-Methylhippuric acid
CAS Number: 27115-49-7
MDL Number: MFCD00044399
Molecular Formula: C10H11NO3
Molecular Weight: 193.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 138
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-(3-Methylbenzoyl)glycine, 3-Methylhippuric Acid
Formula:
C₁₀H₁₁NO₃ Melting Pt: 138 °C |
MDL Number:
MFCD00044399 CAS Number: 27115-49-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...