o-Toluidine-4-sulfonic acid sodium salt tetrahydrate ≥98.0%

Supplier: TCI America

Synonyms: Sodium 4-Amino-3-methylbenzenesulfonate, 2-Aminotoluene-5-sulphonic acid Sodium Salt, Sodium 4-Amino-3-methylbenzenesulphonate, 2-Aminotoluene-5-sulfonic acid Sodium Salt, 4-Amino-3-methylbenzenesulphonic acid Sodium Salt, Sodium 2-Aminotoluene-5-sulphonate, Sodium o-Toluidine-4-sulphonate, Sodium o-Toluidine-4-sulfonate, Sodium 2-Aminotoluene-5-sulfonate, 4-Amino-3-methylbenzenesulfonic acid Sodium Salt

T0304-25G T0304-500G
TCT0304-025GEA 48.98 CAD
TCT0304-025G TCT0304-500G
o-Toluidine-4-sulfonic acid sodium salt tetrahydrate ≥98.0%
o-Toluidine-4-sulfonic acid sodium salt tetrahydrate

CAS Number: 63450-43-1
MDL Number: MFCD00070549
Molecular Formula: C7H9NO3S
Molecular Weight: 209.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Very Pale Yellow


Other Product Names / Synonyms: 2-Aminotoluene-5-sulfonic Acid Sodium Salt, Sodium 2-Aminotoluene-5-sulfonate, Sodium o-Toluidine-4-sulfonate, 4-Amino-3-methylbenzenesulfonic Acid Sodium Salt, Sodium 4-Amino-3-methylbenzenesulfonate

Formula: C₇H₉NO₃S
MW: 209.19 g/mol
MDL Number: MFCD00070549
CAS Number: 63450-43-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8acb0cf57ee5770f3d2b642f76f607559092b17&vid=a8acb0cf57ee5770f3d2b642f76f607559092b17&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR