(Toluene-3,4-dithiolato)zinc(II) ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Zinc Dithiol

T0279-1G T0279-5G
TCT0279-001GEA 121.66 CAD
TCT0279-001G TCT0279-005G
(Toluene-3,4-dithiolato)zinc(II) ≥95.0% (by titrimetric analysis)
(Toluene-3,4-dithiolato)zinc(II)

CAS Number: 29726-21-4
MDL Number: MFCD00005863
Molecular Formula: C7H8S2
Molecular Weight: 219.63
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White


Product Categories: Materials Chemistry, Molecular Conductors
Other Product Names / Synonyms: Zinc Dithiol

Formula: C₇H₈S₂
MW: 219.63 g/mol
MDL Number: MFCD00005863
CAS Number: 29726-21-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41ce356c6eab2b9f2e896f80a2b2ec377fcdb818&vid=41ce356c6eab2b9f2e896f80a2b2ec377fcdb818&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR