Nickel(II) p-toluenesulfonate hexahydrate 98,0 Titrimetric
Supplier: TCI America
Synonyms:
p-Toluenesulphonic acid Nickel(II) Salt, p-Toluenesulfonic acid Nickel(II) Salt
T0276-25G
T0276-5G
TCT0276-025GEA
328.96
CAD
TCT0276-025G
TCT0276-5G
Nickel(II) p-toluenesulfonate hexahydrate 98,0 Titrimetric
Nickel(II) p-toluenesulfonate hexahydrate
CAS Number: 6944-05-4
MDL Number: MFCD00070542
Molecular Formula: C7H8O3S
Molecular Weight: 401.07
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Very Pale Yellow
Product Categories: Synthetic Organic Chemistry, Activation Reaction of Unactivated Bonds
Other Product Names / Synonyms: p-Toluenesulfonic Acid Nickel(II) Salt
Formula:
C₇H₈O₃S MW: 401.07 g/mol |
MDL Number:
MFCD00070542 CAS Number: 6944-05-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...