Sodium toluene-4-sulfonate ≥90.0% for synthesis

Supplier: TCI America

Synonyms: p-Toluenesulphonic acid sodium salt, p-Toluenesulfonic acid sodium salt, Sodium p-toluenesulfonate, Sodium p-toluenesulphonate, Toluene-4-sulfonic acid sodium salt, Toluene-4-sulphonic acid sodium salt

T0271-25G T0271-500G
TCT0271-025GEA 44.24 CAD
TCT0271-025G TCT0271-500G
Sodium toluene-4-sulfonate ≥90.0% for synthesis
Sodium toluene-4-sulphonate

CAS Number: 657-84-1
MDL Number: MFCD00064388
Molecular Formula: C7H8O3S
Molecular Weight: 194.18
Purity/Analysis Method: >90.0% (T)
Form: Crystal
Color: White
Flash Point (°C): 500


Other Product Names / Synonyms: p-Toluenesulfonic Acid Sodium Salt

Formula: C₇H₇NaO₃S
Flash Pt: 500 °C
MDL Number: MFCD00798566
CAS Number: 657-84-1
EINECS: 211-522-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c945c92c8cffa1641a88ebec6008d489da7df3e&vid=9c945c92c8cffa1641a88ebec6008d489da7df3e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR