Titanyl(IV) acetylacetonate

Supplier: TCI America

Synonyms: Titanium(IV) oxide acetylacetonate, Titanium(IV) oxyacetylacetonate, Titanyl Acetylacetonate, Titanium (IV) oxide bis(acetylacetonate), Oxobis(2,4-pentanedionato-O,O')titanium (IV), Titanium (IV) oxide bis(2,4-pentanedionate)

T0249-500G T0249-25G
TCT0249-500GEA 1550.45 CAD
TCT0249-500G TCT0249-025G
Titanyl(IV) acetylacetonate
Titanyl (IV) acetylacetonate

CAS Number: 14024-64-7
MDL Number: MFCD00013505
Molecular Formula: C10H14O5Ti
Molecular Weight: 262.08
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 200


Other Product Names / Synonyms: Acetylacetone Titanium(IV)oxy Salt, Titanium(IV)oxy Acetylacetonate

Formula: C₁₀H₁₄O₅Ti
MW: 262.1 g/mol
Melting Pt: 200 °C
MDL Number: MFCD00013505
CAS Number: 14024-64-7
EINECS: 237-861-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15766d541ae16499b746cb1c142c01137727bbce&vid=15766d541ae16499b746cb1c142c01137727bbce&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR