Dioctadecyl-3,3'-thiodipropionate ≥90.0% (by HPLC)

Supplier: TCI America

T0206-25G T0206-500G
TCT0206-025GEA 62.68 CAD
TCT0206-025G TCT0206-500G
Dioctadecyl-3,3'-thiodipropionate ≥90.0% (by HPLC)
Dioctadecyl-3,3'-thiodipropionate

CAS Number: 693-36-7
MDL Number: MFCD00026684
Molecular Formula: C42H82O4S
Molecular Weight: 683.17
Purity/Analysis Method: >90.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 65
Lambda max.: 410 nm (H2O)


Other Product Names / Synonyms: Distearyl 3,3'-Thiodipropionate, 3,3'-Thiodipropionic Acid Dioctadecyl Ester

Formula: C₄₂H₈₂O₄S
Melting Pt: 65 °C
MDL Number: MFCD00026684
CAS Number: 693-36-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8078a169a9e8d08561a20c681558f022e776ff2&vid=a8078a169a9e8d08561a20c681558f022e776ff2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR