Didodecyl-3,3'-thiodipropionate ≥90.0%

Supplier: TCI America

Synonyms: Dilauryl 3,3'-Thiodipropionate, 3,3'-Thiodipropionic Acid Didodecyl Ester, Dilauryl thiodipropionate

T0205-500G T0205-25G
TCT0205-500GEA 136.46 CAD
TCT0205-500G TCT0205-025G
Didodecyl-3,3'-thiodipropionate ≥90.0%
Didodecyl-3,3'-thiodipropionate

CAS Number: 123-28-4
MDL Number: MFCD00026589
Molecular Formula: C30H58O4S
Molecular Weight: 514.85
Purity/Analysis Method: >90.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 42
Flash Point (°C): 210


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: 3,3'-Thiodipropionic Acid Didodecyl Ester, Dilauryl 3,3'-Thiodipropionate

Formula: C₃₀H₅₈O₄S
Melting Pt: 42 °C
Flash Pt: 210 °C
MDL Number: MFCD00026589
CAS Number: 123-28-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6dd5b174088ec2d14c0de2e4f5c1cfad10d77d88&vid=6dd5b174088ec2d14c0de2e4f5c1cfad10d77d88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR