6,6'-Di-tert-butyl-4,4'-thiodi-m-cresol ≥98.0%
Supplier: TCI America
Synonyms:
4,4'-Thiobis(6-tert-butyl-m-cresol), 4,4'-Thiobis(2-tert-butyl-5-methylphenol), ST029248, Bis(5-tert-butyl-4-hydroxy-2-methylphenyl) sulphide, 4,4'-Thiobis(2-(tert-butyl)-5-methylphenol), Bis(5-tert-butyl-4-hydroxy-2-methylphenyl) sulfide
CAS Number: 96-69-5
MDL Number: MFCD00026287
Molecular Formula: C22H30O2S
Molecular Weight: 358.54
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 162
Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Bis(5-tert-butyl-4-hydroxy-2-methylphenyl) Sulfide
Formula:
C₂₂H₃₀O₂S MW: 358.5374 g/mol Melting Pt: 162 °C |
MDL Number:
MFCD00026287 CAS Number: 96-69-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...