Thiamine nitrate ≥98.0% (by HPLC and UV method)

Supplier: TCI America

T0182-25G T0182-500G T0182-100G
TCT0182-025GEA 133.45 CAD
TCT0182-025G TCT0182-500G TCT0182-100G
Thiamine nitrate ≥98.0% (by HPLC and UV method)
Thiamine nitrate

CAS Number: 532-43-4
MDL Number: MFCD00036330
Molecular Formula: C12H17N5O4S
Molecular Weight: 327.36
Purity/Analysis Method: >98.0% (HPLC,E)
Form: Crystal
Color: White
Melting point (°C): 190


Product Categories: Bioscience, Vitamins
Other Product Names / Synonyms: Vitamin B1 Nitrate

Formula: C₁₂H₁₇N₅O₄S
Melting Pt: 190 °C
MDL Number: MFCD00036330
CAS Number: 532-43-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cb493f3d3d678bd72b4530bf77863fc9cd200ed&vid=3cb493f3d3d678bd72b4530bf77863fc9cd200ed&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR