1,3,5-Triphenylformazan ≥92.0%

Supplier: TCI America

Synonyms: 1,3,5-Triphenyl tetrazolium formazan

T0175-1G T0175-25G
TCT0175-001GEA 48.21 CAD
TCT0175-001G TCT0175-025G
1,3,5-Triphenylformazan ≥92.0%
1,3,5-Triphenylformazan

CAS Number: 531-52-2
MDL Number: MFCD00020647
Molecular Formula: C19H16N4
Molecular Weight: 300.37
Purity/Analysis Method: >92.0% (HPLC)
Form: Crystal
Color: Yellow Red


Other Product Names / Synonyms: Tetrazole Red Formazan, Tetrazolium Red Formazan, TTC Formazan

Formula: C₁₉H₁₆N₄
MDL Number: MFCD00020647
CAS Number: 531-52-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5983be39ba4cf89aa559340b82478aa6e2875fcb&vid=5983be39ba4cf89aa559340b82478aa6e2875fcb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 92.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR