Tetrapropenylsuccinic anhydride (mixture of branched chain isomers) ≥85.0% (by titrimetric analysis)

Supplier: TCI America

T0170-500G T0170-25G
TCT0170-500GEA 194.34 CAD
TCT0170-500G TCT0170-025G
Tetrapropenylsuccinic anhydride (mixture of branched chain isomers) ≥85.0% (by titrimetric analysis)
Tetrapropenylsuccinic anhydride (mixture of branched chain isomers)

CAS Number: 26544-38-7
MDL Number: MFCD00005528
Molecular Formula: C16H26O3
Molecular Weight: 266.38
Purity/Analysis Method: >85.0% (T)
Form: Clear Liquid
Color: Pale Yellow
Flash Point (°C): 185
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: Dodecenylsuccinic Anhydride, Isododecenylsuccinic Anhydride

Formula: C₁₆H₂₆O₃
Density: 1.01 (20 °C)
Flash Pt: 185 °C
MDL Number: MFCD00005528
CAS Number: 26544-38-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38d71a0ac6d44acaec8d1a261a6d319d7773dff0&vid=38d71a0ac6d44acaec8d1a261a6d319d7773dff0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR