3',3'',5',5''-Tetraiodophenolphthalein ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Tetraiodophthalein, Iodophene, Iodophthalein

T0126-25G T0126-5G
TCT0126-025GEA 922.72 CAD
TCT0126-025G TCT0126-5G
3',3'',5',5''-Tetraiodophenolphthalein ≥95.0% (by HPLC, titration analysis)
3',3'',5',5''-Tetraiodophenolphthalein

CAS Number: 386-17-4
MDL Number: MFCD00001614
Molecular Formula: C20H10I4O4
Molecular Weight: 821.91
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Color: Very Pale Yellow


Other Product Names / Synonyms: Iodophene, Iodophthalein, Tetraiodophthalein

Formula: C₂₀H₁₀I₄O₄
MW: 821.91 g/mol
MDL Number: MFCD00001614
CAS Number: 386-17-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31a7ff695bdc5c9964a05846c7cdfa5ae44b180c&vid=31a7ff695bdc5c9964a05846c7cdfa5ae44b180c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR