Tetrahydroxy-1,4-benzoquinone disodium salt

Supplier: TCI America

Synonyms: Sodium 4,5-Dihydroxy-3,6-dioxocyclohexa-1,4-diene-1,2-bis(olate), Tetrahydroxyquinone Disodium Salt

T0122-25G T0122-1G
TCT0122-025GEA 273.34 CAD
TCT0122-025G TCT0122-001G
Tetrahydroxy-1,4-benzoquinone disodium salt
Tetrahydroxy-1,4-benzoquinone disodium salt

CAS Number: 1887-02-1
MDL Number: MFCD00036127
Molecular Formula: C6H4O6
Molecular Weight: 216.06
Form: Crystal
Color: Yellow Green


Other Product Names / Synonyms: Tetrahydroxyquinone Disodium Salt

Formula: C₆H₄O₆
MW: 216.06 g/mol
MDL Number: MFCD00036127
CAS Number: 1887-02-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46bb2839b6fa0ebf318e900b262efbe75e0def19&vid=46bb2839b6fa0ebf318e900b262efbe75e0def19&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR