Diglycerol mixture of isomers ≥80.0%

Supplier: TCI America

T0119-25G T0119-500G
TCT0119-025GEA 56.88 CAD
TCT0119-025G TCT0119-500G
Diglycerol mixture of isomers ≥80.0%
Diglycerol

CAS Number: 627-82-7
MDL Number: MFCD00049316
Molecular Formula: C6H14O5
Molecular Weight: 166.17
Purity/Analysis Method: >80.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 240
Specific Gravity (20/20): 1.28


Other Product Names / Synonyms: Oxydi(propanediol), Tetrahydroxydipropyl Ether

Formula: C₆H₁₄O₅
Density: 1.28 (20 °C)
Flash Pt: 240 °C
MDL Number: MFCD00049316
CAS Number: 627-82-7

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR