Leucoquinizarin ≥98.0% (by HPLC)

Supplier: TCI America

T0116-500G T0116-25G
TCT0116-500GEA 906.92 CAD
TCT0116-500G TCT0116-025G
Leucoquinizarin ≥98.0% (by HPLC)
Leucoquinizarin

CAS Number: 476-60-8
MDL Number: MFCD00058948
Molecular Formula: C14H10O4
Molecular Weight: 242.23
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: Yellow
Melting point (°C): 151


Other Product Names / Synonyms: 2,3-Dihydro-9,10-dihydroxy-1,4-anthraquinone, 1,4-Dihydroxyanthrahydroquinone, leuco-Quinizarin, 1,4,9,10-Tetrahydroxyanthracene

Formula: C₁₄H₁₀O₄
Melting Pt: 151 °C
MDL Number: MFCD00058948
CAS Number: 476-60-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e527ca4d9ecb26b716da72390d9c5744007596c&vid=4e527ca4d9ecb26b716da72390d9c5744007596c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR