1,2,3,4-Tetrahydroquinaldine ≥97.0%
Supplier: TCI America
Synonyms:
2-Methyl-1,2,3,4-tetrahydroquinoline, 1,2,3,4-Tetrahydro-2-methylquinoline
T0112-25G
T0112-500G
TCT0112-025GEA
81.97
CAD
TCT0112-025G
TCT0112-500G
1,2,3,4-Tetrahydroquinaldine ≥97.0%
1,2,3,4-Tetrahydroquinaldine
CAS Number: 1780-19-4
MDL Number: MFCD00023886
Molecular Formula: C10H13N
Molecular Weight: 147.22
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 250
Specific Gravity (20/20): 1.02
Other Product Names / Synonyms: 2-Methyl-1,2,3,4-tetrahydroquinoline, 1,2,3,4-Tetrahydro-2-methylquinoline
Formula:
C₁₀H₁₃N MW: 147.22 g/mol Boiling Pt: 250 °C Density: 1.02 (20 °C) |
MDL Number:
MFCD00023886 CAS Number: 1780-19-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...