Tetrachlorophthalic acid hemihydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3,4,5,6-Tetrachlorophthalic acid

T0070-500G T0070-25G
TCT0070-500GEA 287.78 CAD
TCT0070-500G TCT0070-025G
Tetrachlorophthalic acid hemihydrate ≥98.0% (by titrimetric analysis)
Tetrachlorophthalic acid hemihydrate

CAS Number: 632-58-6
MDL Number: MFCD00053309
Molecular Formula: C8H2Cl4O4
Molecular Weight: 303.90
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White

Formula: C₈H₂Cl₄O₄
MW: 303.90 g/mol
MDL Number: MFCD00053309
CAS Number: 632-58-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e26df43f453408a82f5229553a784436491e695&vid=1e26df43f453408a82f5229553a784436491e695&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR