Calcium mesotartrate trihydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Mesotartaric Acid Calcium Salt, meso-Tartaric Acid Calcium Salt, Calcium Mesotartrate
T0004-1G
T0004-25G
TCT0004-001GEA
67.94
CAD
TCT0004-001G
TCT0004-025G
Calcium mesotartrate trihydrate ≥98.0% (by titrimetric analysis)
Calcium mesotartrate trihydrate
CAS Number: 815-79-2
MDL Number: MFCD00151483
Molecular Formula: C4H6O6
Molecular Weight: 188.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Other Product Names / Synonyms: Calcium Mesotartrate, Mesotartaric Acid Calcium Salt, meso-Tartaric Acid Calcium Salt
Formula:
C₄H₆O₆ MW: 188.15 g/mol |
MDL Number:
MFCD00151483 CAS Number: 815-79-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...