Silver(II) picolinate ≥97.0%

Supplier: TCI America

S0815-1G S0815-5G
TCS0815-1GEA 118.5 CAD
TCS0815-1G TCS0815-5G
Silver(II) picolinate ≥97.0%
Silver(II) picolinate

CAS Number: 14783-00-7
Molecular Formula: C12H8AgN2O4
Molecular Weight: 352.07
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: Pale Yellow
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Oxidation
Other Product Names / Synonyms: Picolinic Acid Silver(II) Salt, Silver(II) Picolinate, Pyridine-2-carboxylic Acid Silver(II) Salt

Formula: C₁₂H₈AgN₂O₄
MDL Number: MFCD15072194
CAS Number: 14783-00-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81440a2d8c5a62b6a2068f07d6a7957ec4e0bd87&vid=81440a2d8c5a62b6a2068f07d6a7957ec4e0bd87&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR