N-Succinimidyl Acrylate ≥98.0%

Supplier: TCI America

Synonyms: N-Acryloxysuccinimide, 2,5-Dioxopyrrolidin-1-yl acrylate, N-Acryloyloxysuccinimide, Acrylic Acid N-Succinimidyl Ester

S0814-5G S0814-25G
TCS0814-5GEA 295.93 CAD
TCS0814-5G TCS0814-25G
N-Succinimidyl Acrylate ≥98.0%
N-Succinimidyl Acrylate

CAS Number: 38862-24-7
MDL Number: MFCD00078261
Molecular Formula: C7H7NO4
Molecular Weight: 169.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 71
Storage Temperature: <0°C


Product Categories: Materials Chemistry, Monomers, Bioscience, Crosslinkers
Other Product Names / Synonyms: Acrylic Acid N-Succinimidyl Ester, N-Acryloyloxysuccinimide

Formula: C₇H₇NO₄
MW: 169.14 g/mol
Melting Pt: 71 °C
MDL Number: MFCD00078261
CAS Number: 38862-24-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7c7f952c70f918d608f402c4380b2c9af66ffd4&vid=c7c7f952c70f918d608f402c4380b2c9af66ffd4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR